Assay Details
| Gene Symbol: AT3G57350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115595.7 | NP_191294.5 | 6 - 7 | 785 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G57350| Gene Symbol: AT3G57350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115595.7 | NP_191294.5 | 6 - 7 | 785 | 95 | |
Set Membership: |
|