Assay Details
| Gene Symbol: ADSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115621.5 | NP_191320.1 | 3 - 4 | 1416 | 64 | |
| GenBank mRNA | AY054606.1 | - | 3 - 4 | 1354 | 64 | |
| AY081461.1 | - | 3 - 4 | 1299 | 64 | ||
| U49389.1 | - | 3 - 4 | 1332 | 64 | ||
More Information
Set Membership: |
|

ADSS
Genomic Map