Assay Details
| Gene Symbol: LPAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115625.4 | NP_567052.1 | 3 - 4 | 616 | 97 | |
| GenBank mRNA | BT026036.1 | - | 3 - 4 | 248 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LPAT2| Gene Symbol: LPAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115625.4 | NP_567052.1 | 3 - 4 | 616 | 97 | |
| GenBank mRNA | BT026036.1 | - | 3 - 4 | 248 | 97 | |
Set Membership: |
|