Assay Details
| Gene Symbol: LPAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115625.4 | NP_567052.1 | 4 - 5 | 685 | 100 | |
| GenBank mRNA | BT026036.1 | - | 4 - 5 | 317 | 100 | |
More Information
Set Membership: |
|

LPAT2
Genomic Map