Assay Details
| Gene Symbol: AGAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036778.2 | NP_001031855.1 | 14 - 15 | 1133 | 112 | |
| NM_120921.5 | NP_568193.1 | 13 - 14 | 1246 | 112 | ||
| GenBank mRNA | AK317201.1 | - | 14 - 15 | 1134 | 112 | |
| AY085529.1 | - | 13 - 14 | 1162 | 112 | ||
| AY090238.1 | - | 13 - 14 | 1113 | 112 | ||
| BT000619.1 | - | 13 - 14 | 1102 | 112 | ||
More Information
Set Membership: |
|

AGAL2
Genomic Map