Assay Details
| Gene Symbol: AGAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036778.2 | NP_001031855.1 | 4 - 5 | 278 | 93 | |
| NM_120921.5 | NP_568193.1 | 3 - 4 | 391 | 93 | ||
| GenBank mRNA | AK317201.1 | - | 4 - 5 | 279 | 93 | |
| AY085529.1 | - | 3 - 4 | 307 | 93 | ||
| AY090238.1 | - | 3 - 4 | 258 | 93 | ||
| BT000619.1 | - | 3 - 4 | 247 | 93 | ||
More Information
Set Membership: |
|

AGAL2
Genomic Map