Assay Details
| Gene Symbol: AGAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342998.1 | NP_001332077.1 | 13 - 14 | 1337 | 70 | |
| NM_120922.5 | NP_196455.1 | 14 - 15 | 1306 | 70 | ||
More Information
Set Membership: |
|

AGAL1
Genomic Map