Assay Details
| Gene Symbol: AT5G08415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120926.3 | NP_568196.1 | 5 - 6 | 1147 | 98 | |
| GenBank mRNA | AB073745.1 | - | 5 - 6 | 1013 | 98 | |
| BT025549.1 | - | 5 - 6 | 993 | 98 | ||
More Information
Set Membership: |
|

AT5G08415
Genomic Map