Assay Details
| Gene Symbol: ILP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120941.2 | NP_196472.1 | 3 - 4 | 1965 | 93 | |
| GenBank mRNA | AB253763.1 | - | 3 - 4 | 1965 | 93 | |
More Information
Set Membership: |
|

ILP1
Genomic Map