Assay Details
| Gene Symbol: AT5G09570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120994.3 | NP_196519.1 | 4 - 5 | 561 | 78 | |
| GenBank mRNA | AK118694.1 | - | 4 - 5 | 390 | 78 | |
| BT003661.1 | - | 4 - 5 | 326 | 78 | ||
More Information
Set Membership: |
|

AT5G09570
Genomic Map