Assay Details
| Gene Symbol: MTHSC70-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120996.4 | NP_196521.1 | 1 - 2 | 359 | 77 | |
| GenBank mRNA | AF217458.1 | - | 1 - 2 | 132 | 77 | |
| BT046197.1 | - | 1 - 2 | 74 | 77 | ||
More Information
Set Membership: |
|

MTHSC70-2
Genomic Map