Assay Details
| Gene Symbol: JAZ10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161241.2 | NP_001154713.1 | 1 - 2 | 281 | 114 | |
| NM_001343260.1 | NP_001330992.1 | 1 - 2 | 323 | 114 | ||
| NM_001343261.1 | NP_001330993.1 | 1 - 2 | 281 | 114 | ||
| NM_001343262.1 | NP_001330994.1 | 1 - 2 | 281 | 114 | ||
| NM_121325.3 | NP_568287.1 | 1 - 2 | 323 | 114 | ||
| NM_203046.3 | NP_974775.1 | 1 - 2 | 323 | 114 | ||
| NM_203047.3 | NP_974776.1 | 1 - 2 | 323 | 114 | ||
| GenBank mRNA | AY056444.1 | - | 1 - 2 | 192 | 114 | |
| AY081715.1 | - | 1 - 2 | 108 | 114 | ||
| FJ417331.1 | - | 1 - 2 | 108 | 114 | ||
More Information
Set Membership: |
|

JAZ10
Genomic Map