Assay Details
| Gene Symbol: JAZ10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343262.1 | NP_001330994.1 | 2 - 3 | 776 | 80 | |
| NM_121325.3 | NP_568287.1 | 3 - 4 | 707 | 80 | ||
| NM_203046.3 | NP_974775.1 | 3 - 4 | 707 | 80 | ||
| NM_203047.3 | NP_974776.1 | 3 - 4 | 707 | 80 | ||
| GenBank mRNA | AY056444.1 | - | 3 - 4 | 576 | 80 | |
| AY081715.1 | - | 3 - 4 | 492 | 80 | ||
More Information
Set Membership: |
|

JAZ10
Genomic Map