Assay Details
| Gene Symbol: DND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121545.5 | NP_197045.1 | 2 - 3 | 821 | 116 | |
| NM_203054.2 | NP_974783.2 | 3 - 4 | 948 | 116 | ||
| GenBank mRNA | AF067798.1 | - | 2 - 3 | 756 | 116 | |
More Information
Set Membership: |
|

DND1
Genomic Map