Assay Details
| Gene Symbol: DND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121545.5 | NP_197045.1 | 6 - 7 | 1702 | 79 | |
| NM_203054.2 | NP_974783.2 | 7 - 8 | 1829 | 79 | ||
| GenBank mRNA | AF067798.1 | - | 6 - 7 | 1637 | 79 | |
More Information
Set Membership: |
|

DND1
Genomic Map