Assay Details
| Gene Symbol: CAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121644.4 | NP_197143.1 | 1 - 2 | 429 | 68 | |
| NM_203060.2 | NP_974789.1 | 1 - 2 | 258 | 68 | ||
| GenBank mRNA | AF236873.1 | - | 1 - 2 | 244 | 68 | |
| AY054617.1 | - | 1 - 2 | 255 | 68 | ||
| AY072460.1 | - | 1 - 2 | 152 | 68 | ||
More Information
Set Membership: |
|

CAC1
Genomic Map