Assay Details
| Gene Symbol: CAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121644.4 | NP_197143.1 | 3 - 4 | 632 | 115 | |
| NM_203060.2 | NP_974789.1 | 3 - 4 | 461 | 115 | ||
| GenBank mRNA | AF236873.1 | - | 3 - 4 | 447 | 115 | |
| AY054617.1 | - | 3 - 4 | 458 | 115 | ||
| AY072460.1 | - | 3 - 4 | 355 | 115 | ||
| AY087857.1 | - | 3 - 4 | 461 | 115 | ||
More Information
Set Membership: |
|

CAC1
Genomic Map