Assay Details
| Gene Symbol: PIR121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125776.1 | NP_001119248.1 | 11 - 12 | 1060 | 87 | |
| NM_121846.4 | NP_197342.3 | 11 - 12 | 1009 | 87 | ||
| NM_203072.3 | NP_974801.2 | 11 - 12 | 1133 | 87 | ||
| GenBank mRNA | AY496701.1 | - | 11 - 12 | 1060 | 87 | |
| AY662957.1 | - | 10 - 11 | 886 | 87 | ||
| AY787212.1 | - | 10 - 11 | 886 | 87 | ||
More Information
Set Membership: |
|

PIR121
Genomic Map