Assay Details
| Gene Symbol: PIR121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125776.1 | NP_001119248.1 | 15 - 16 | 1723 | 67 | |
| NM_121846.4 | NP_197342.3 | 15 - 16 | 1672 | 67 | ||
| NM_203072.3 | NP_974801.2 | 15 - 16 | 1796 | 67 | ||
| GenBank mRNA | AY496701.1 | - | 15 - 16 | 1723 | 67 | |
| AY662957.1 | - | 14 - 15 | 1549 | 67 | ||
| AY787212.1 | - | 14 - 15 | 1549 | 67 | ||
More Information
Set Membership: |
|

PIR121
Genomic Map