Assay Details
| Gene Symbol: EIF2 BETA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085141.1 | NP_001078610.1 | 5 - 6 | 472 | 93 | |
| NM_122100.4 | NP_197592.1 | 5 - 6 | 402 | 93 | ||
| NM_203088.2 | NP_974817.1 | 5 - 6 | 365 | 93 | ||
| GenBank mRNA | AY128324.1 | - | 5 - 6 | 331 | 93 | |
| BT000056.1 | - | 4 - 5 | 260 | 93 | ||
More Information
Set Membership: |
|

EIF2 BETA
Genomic Map