Assay Details
| Gene Symbol: CDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343786.1 | NP_001331796.1 | 3 - 4 | 268 | 58 | |
| NM_122210.5 | NP_197695.1 | 2 - 3 | 293 | 58 | ||
| GenBank mRNA | AB210817.1 | - | 2 - 3 | 218 | 58 | |
| AY125520.1 | - | 2 - 3 | 218 | 58 | ||
| AY143835.1 | - | 2 - 3 | 168 | 58 | ||
More Information
Set Membership: |
|

CDF1
Genomic Map