Assay Details
| Gene Symbol: CDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343786.1 | NP_001331796.1 | 5 - 6 | 732 | 105 | |
| NM_122210.5 | NP_197695.1 | 4 - 5 | 757 | 105 | ||
| GenBank mRNA | AB210817.1 | - | 4 - 5 | 682 | 105 | |
| AY125520.1 | - | 4 - 5 | 682 | 105 | ||
| AY143835.1 | - | 4 - 5 | 632 | 105 | ||
More Information
Set Membership: |
|

CDF1
Genomic Map