Assay Details
| Gene Symbol: DES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344053.1 | NP_001330586.1 | 4 - 5 | 746 | 80 | |
| NM_001344054.1 | NP_001330588.1 | 4 - 5 | 765 | 80 | ||
| NM_001344055.1 | NP_001330587.1 | 4 - 5 | 667 | 80 | ||
| NM_122686.2 | NP_198155.1 | 3 - 4 | 627 | 80 | ||
| NM_203114.2 | NP_974843.1 | 4 - 5 | 815 | 80 | ||
More Information
Set Membership: |
|

DES1
Genomic Map