Assay Details
| Gene Symbol: DES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344054.1 | NP_001330588.1 | 7 - 8 | 1037 | 122 | |
| NM_001344055.1 | NP_001330587.1 | 7 - 8 | 939 | 122 | ||
| NM_122686.2 | NP_198155.1 | 6 - 7 | 899 | 122 | ||
| NM_203114.2 | NP_974843.1 | 7 - 8 | 1087 | 122 | ||
More Information
Set Membership: |
|

DES1
Genomic Map