Assay Details
| Gene Symbol: MBD02 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203492.2 | NP_001190421.1 | 2 - 3 | 621 | 66 | |
| NM_001344102.1 | NP_001331130.1 | 1 - 2 | 1001 | 66 | ||
| NM_122924.3 | NP_198383.1 | 2 - 3 | 640 | 66 | ||
| NM_203121.3 | NP_974850.1 | 2 - 3 | 661 | 66 | ||
| GenBank mRNA | AY088026.1 | - | 2 - 3 | 593 | 66 | |
| BT025800.1 | - | 1 - 2 | 438 | 66 | ||
More Information
Set Membership: |
|

MBD02
Genomic Map