Assay Details
| Gene Symbol: AT5G42850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123650.3 | NP_568614.1 | 1 - 2 | 320 | 89 | |
| NM_203143.2 | NP_974872.1 | 1 - 2 | 199 | 89 | ||
| GenBank mRNA | AK118124.1 | - | 1 - 2 | 200 | 89 | |
| BT005444.1 | - | 1 - 2 | 145 | 89 | ||
More Information
Set Membership: |
|

AT5G42850
Genomic Map