Assay Details
| Gene Symbol: PAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344761.1 | NP_001332482.1 | 3 - 4 | 1410 | 82 | |
| NM_001344762.1 | NP_001332483.1 | 2 - 3 | 1445 | 82 | ||
| NM_124189.6 | NP_199626.1 | 2 - 3 | 1341 | 82 | ||
| NM_203174.3 | NP_974903.1 | 3 - 4 | 1421 | 82 | ||
| GenBank mRNA | AF153443.1 | - | 2 - 3 | 1154 | 82 | |
| AK317252.1 | - | 2 - 3 | 1209 | 82 | ||
| AK317747.1 | - | 3 - 4 | 1251 | 82 | ||
| BT025326.1 | - | 1 - 2 | 1025 | 82 | ||
More Information
Set Membership: |
|

PAT1
Genomic Map