Assay Details
| Gene Symbol: AGL71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203588.1 | NP_001190517.2 | 1 - 2 | 270 | 97 | |
| NM_001344965.1 | NP_001331386.1 | 2 - 3 | 263 | 97 | ||
| NM_001344966.1 | NP_001331387.1 | 2 - 3 | 278 | 97 | ||
| NM_124566.3 | NP_200000.3 | 1 - 2 | 189 | 97 | ||
| NM_203195.1 | NP_974924.1 | 1 - 2 | 189 | 97 | ||
| GenBank mRNA | AY141220.1 | - | 1 - 2 | 189 | 97 | |
More Information
Set Membership: |
|

AGL71
Genomic Map