Assay Details
| Gene Symbol: AGL71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203588.1 | NP_001190517.2 | 2 - 3 | 349 | 100 | |
| NM_001344965.1 | NP_001331386.1 | 3 - 4 | 342 | 100 | ||
| NM_001344966.1 | NP_001331387.1 | 3 - 4 | 357 | 100 | ||
| NM_124566.3 | NP_200000.3 | 2 - 3 | 268 | 100 | ||
| NM_203195.1 | NP_974924.1 | 2 - 3 | 268 | 100 | ||
| GenBank mRNA | AY141220.1 | - | 2 - 3 | 268 | 100 | |
More Information
Set Membership: |
|

AGL71
Genomic Map