Assay Details
| Gene Symbol: AT5G53850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345068.1 | NP_001330484.1 | 5 - 6 | 1109 | 107 | |
| NM_001345069.1 | NP_001330483.1 | 5 - 6 | 822 | 107 | ||
| NM_001345070.1 | NP_001330482.1 | 4 - 5 | 1198 | 107 | ||
| NM_124764.2 | NP_200196.2 | 4 - 5 | 651 | 107 | ||
| NM_203202.3 | NP_974931.1 | 4 - 5 | 894 | 107 | ||
| NM_203203.2 | NP_974932.1 | 5 - 6 | 765 | 107 | ||
| GenBank mRNA | BT012606.1 | - | 4 - 5 | 627 | 107 | |
| BT015100.1 | - | 4 - 5 | 561 | 107 | ||
More Information
Set Membership: |
|

AT5G53850
Genomic Map