Assay Details
| Gene Symbol: UCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125287.5 | NP_568894.1 | 1 - 2 | 348 | 67 | |
| NM_203233.1 | NP_974962.1 | 1 - 2 | 314 | 67 | ||
| GenBank mRNA | AB021706.1 | - | 1 - 2 | 89 | 67 | |
| AY088200.1 | - | 1 - 2 | 245 | 67 | ||
| JQ937241.1 | - | 1 - 2 | 74 | 67 | ||
More Information
Set Membership: |
|

UCP2
Genomic Map