Assay Details
| Gene Symbol: BON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125583.4 | NP_568944.1 | 3 - 4 | 282 | 142 | |
| NM_203248.2 | NP_974977.1 | 2 - 3 | 317 | 142 | ||
| GenBank mRNA | AX380787.1 | - | 2 - 3 | 239 | 142 | |
| AY062824.1 | - | 3 - 4 | 273 | 142 | ||
| BT010555.1 | - | 2 - 3 | 195 | 142 | ||
| Gene Symbol: AT5G61910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203661.1 | NP_001190590.1 | 9 - 10 | 2718 | 142 | |
More Information
Set Membership: |
|

BON1
Genomic Map