Assay Details
| Gene Symbol: BON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125583.4 | NP_568944.1 | 6 - 7 | 545 | 66 | |
| NM_203248.2 | NP_974977.1 | 5 - 6 | 580 | 66 | ||
| GenBank mRNA | AX380787.1 | - | 5 - 6 | 502 | 66 | |
| AY062824.1 | - | 6 - 7 | 536 | 66 | ||
| AY085339.1 | - | 6 - 7 | 537 | 66 | ||
| BT010555.1 | - | 5 - 6 | 458 | 66 | ||
| Gene Symbol: AT5G61910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203661.1 | NP_001190590.1 | 12 - 13 | 2981 | 66 | |
More Information
Set Membership: |
|

BON1
Genomic Map