Assay Details
| Gene Symbol: BON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125583.4 | NP_568944.1 | 7 - 8 | 730 | 145 | |
| NM_203248.2 | NP_974977.1 | 6 - 7 | 765 | 145 | ||
| GenBank mRNA | AX380787.1 | - | 6 - 7 | 687 | 145 | |
| AY062824.1 | - | 7 - 8 | 721 | 145 | ||
| AY085339.1 | - | 7 - 8 | 722 | 145 | ||
| BT010555.1 | - | 6 - 7 | 643 | 145 | ||
| Gene Symbol: AT5G61910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203661.1 | NP_001190590.1 | 13 - 14 | 3166 | 145 | |
More Information
Set Membership: |
|

BON1
Genomic Map