Assay Details
| Gene Symbol: BON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125583.4 | NP_568944.1 | 8 - 9 | 863 | 86 | |
| NM_203248.2 | NP_974977.1 | 7 - 8 | 898 | 86 | ||
| GenBank mRNA | AX380787.1 | - | 7 - 8 | 820 | 86 | |
| AY062824.1 | - | 8 - 9 | 854 | 86 | ||
| AY085339.1 | - | 8 - 9 | 855 | 86 | ||
| BT010555.1 | - | 7 - 8 | 776 | 86 | ||
| Gene Symbol: AT5G61910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203661.1 | NP_001190590.1 | 14 - 15 | 3299 | 86 | |
More Information
Set Membership: |
|

BON1
Genomic Map