Assay Details
| Gene Symbol: NAC012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103011.2 | NP_174554.1 | 1 - 2 | 389 | 150 | |
| GenBank mRNA | EF101892.1 | - | 1 - 2 | 185 | 150 | |
More Information
Set Membership: |
|

NAC012
Genomic Map