Assay Details
| Gene Symbol: NAC012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103011.2 | NP_174554.1 | 2 - 3 | 655 | 104 | |
| GenBank mRNA | EF101892.1 | - | 2 - 3 | 451 | 104 | |
More Information
Set Membership: |
|

NAC012
Genomic Map