Assay Details
| Gene Symbol: AT1G32880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103022.1 | NP_174565.1 | 1 - 2 | 126 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G32880| Gene Symbol: AT1G32880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103022.1 | NP_174565.1 | 1 - 2 | 126 | 99 | |
Set Membership: |
|