Assay Details
| Gene Symbol: GBSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103023.4 | NP_174566.1 | 12 - 13 | 1989 | 65 | |
| GenBank mRNA | AY088544.1 | - | 12 - 13 | 1871 | 65 | |
| AY094405.1 | - | 12 - 13 | 1791 | 65 | ||
| AY123983.1 | - | 12 - 13 | 1889 | 65 | ||
| AY149948.1 | - | 11 - 12 | 1722 | 65 | ||
| KJ138976.1 | - | 11 - 12 | 1722 | 65 | ||
More Information
Set Membership: |
|

GBSS1
Genomic Map