Assay Details
| Gene Symbol: GBSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103023.4 | NP_174566.1 | 7 - 8 | 1043 | 64 | |
| GenBank mRNA | AY088544.1 | - | 7 - 8 | 925 | 64 | |
| AY094405.1 | - | 7 - 8 | 845 | 64 | ||
| AY123983.1 | - | 7 - 8 | 943 | 64 | ||
| AY149948.1 | - | 6 - 7 | 776 | 64 | ||
| KJ138976.1 | - | 6 - 7 | 776 | 64 | ||
More Information
Set Membership: |
|

GBSS1
Genomic Map