Assay Details
| Gene Symbol: GTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333032.1 | NP_001322396.1 | 1 - 2 | 723 | 139 | |
| NM_103052.4 | NP_174594.1 | 1 - 2 | 723 | 139 | ||
More Information
Set Membership: |
|

GTL1
Genomic Map