Assay Details
| Gene Symbol: NAC015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103057.4 | NP_174598.1 | 2 - 3 | 559 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAC015| Gene Symbol: NAC015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103057.4 | NP_174598.1 | 2 - 3 | 559 | 106 | |
Set Membership: |
|