Assay Details
| Gene Symbol: AT1G33360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103063.4 | NP_564423.3 | 1 - 2 | 1142 | 102 | |
| GenBank mRNA | BT015363.1 | - | 1 - 2 | 997 | 102 | |
| BT015893.1 | - | 1 - 2 | 811 | 102 | ||
More Information
Set Membership: |
|

AT1G33360
Genomic Map