Assay Details
| Gene Symbol: AT1G33440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103069.4 | NP_174610.2 | 4 - 5 | 1036 | 68 | |
| GenBank mRNA | AK221617.1 | - | 4 - 5 | 999 | 68 | |
More Information
Set Membership: |
|

AT1G33440
Genomic Map