Assay Details
| Gene Symbol: ADR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103079.4 | NP_174620.2 | 2 - 3 | 1293 | 90 | |
| GenBank mRNA | AJ581996.1 | - | 2 - 3 | 969 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADR1| Gene Symbol: ADR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103079.4 | NP_174620.2 | 2 - 3 | 1293 | 90 | |
| GenBank mRNA | AJ581996.1 | - | 2 - 3 | 969 | 90 | |
Set Membership: |
|