Assay Details
| Gene Symbol: ADR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103079.4 | NP_174620.2 | 3 - 4 | 1655 | 88 | |
| GenBank mRNA | AJ581996.1 | - | 3 - 4 | 1331 | 88 | |
More Information
Set Membership: |
|

ADR1
Genomic Map