Assay Details
| Gene Symbol: MES14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103121.4 | NP_174661.1 | 2 - 3 | 845 | 140 | |
| GenBank mRNA | AF424607.1 | - | 2 - 3 | 834 | 140 | |
| AK226919.1 | - | 2 - 3 | 647 | 140 | ||
| BT025240.1 | - | 2 - 3 | 493 | 140 | ||
More Information
Set Membership: |
|

MES14
Genomic Map