Assay Details
| Gene Symbol: AT1G34020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333079.1 | NP_001321552.1 | 5 - 6 | 968 | 114 | |
| NM_001333080.1 | NP_001321551.1 | 4 - 5 | 1210 | 114 | ||
| NM_103124.4 | NP_564433.1 | 5 - 6 | 947 | 114 | ||
| GenBank mRNA | BT033114.1 | - | 4 - 5 | 682 | 114 | |
| KJ667162.1 | - | 4 - 5 | 682 | 114 | ||
| KP872773.1 | - | 4 - 5 | 682 | 114 | ||
More Information
Set Membership: |
|

AT1G34020
Genomic Map