Assay Details
Gene Symbol: FAB1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333092.1 | NP_001319143.1 | 3 - 4 | 375 | 115 | |
NM_001333093.1 | NP_001321920.1 | 3 - 4 | 305 | 115 | ||
NM_103148.4 | NP_174686.1 | 2 - 3 | 118 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
