Assay Details
| Gene Symbol: AT1G35180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103215.5 | NP_174751.2 | 3 - 4 | 778 | 94 | |
| GenBank mRNA | BT010848.1 | - | 3 - 4 | 638 | 94 | |
| BT015379.1 | - | 3 - 4 | 587 | 94 | ||
More Information
Set Membership: |
|

AT1G35180
Genomic Map